The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018266.2 | NP_060736.1 | 5 - 6 | 999 | 68 |
NR_073506.1 | 6 - 7 | 1108 | 68 | ||
XM_005247578.1 | XP_005247635.1 | 5 - 6 | 964 | 68 | |
XM_006713687.1 | XP_006713750.1 | 5 - 6 | 907 | 68 | |
XM_011512958.2 | XP_011511260.1 | 5 - 6 | 964 | 68 | |
XM_017006788.1 | XP_016862277.1 | 4 - 5 | 880 | 68 | |
GenBank mRNA | AK001764.1 | - | 5 - 6 | 941 | 68 |
AK222950.1 | - | 5 - 6 | 931 | 68 | |
AK223248.1 | - | 5 - 6 | 931 | 68 | |
BC003192.1 | - | 6 - 7 | 1023 | 68 | |
BC010070.2 | - | 6 - 7 | 1023 | 68 | |
BC021277.2 | - | 5 - 6 | 917 | 68 |