The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284407.1 | NP_001271336.1 | 1 - 2 | 122 | 61 |
NM_001284408.1 | NP_001271337.1 | 1 - 2 | 122 | 61 | |
NM_018306.3 | NP_060776.2 | 1 - 2 | 122 | 61 | |
GenBank mRNA | AK001898.1 | - | 1 - 2 | 54 | 61 |
AK092470.1 | - | 1 - 2 | 122 | 61 | |
BC020658.1 | - | 1 - 2 | 72 | 61 | |
BG335433.1 | - | 1 - 2 | 61 | 61 |