Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MIS18BP1
Entrez Gene ID
55320
Gene Name
MIS18 binding protein 1
Gene Aliases
C14orf106, HSA242977, KNL2, M18BP1
Chromosome Location
Chr.14: 45203190 - 45253402 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018353.4 NP_060823.3 1 - 2 368 83
GenBank mRNA AB067490.1 - 1 - 2 368 83
AK002048.1 - 1 - 2 164 83
BC051885.1 - 1 - 2 145 83
BC065544.1 - 1 - 2 159 83

View Full Product Details