The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018376.3 | NP_060846.2 | 1 - 2 | 562 | 75 |
NR_130760.1 | 1 - 2 | 562 | 75 | ||
XM_011518839.1 | XP_011517141.1 | 1 - 2 | 562 | 75 | |
XM_017014889.1 | XP_016870378.1 | 1 - 2 | 514 | 75 | |
GenBank mRNA | AK002137.1 | - | 1 - 2 | 167 | 75 |
BC005202.1 | - | 1 - 2 | 95 | 75 | |
BC017914.1 | - | 1 - 2 | 133 | 75 | |
BX647652.1 | - | 1 - 2 | 134 | 75 | |
DA346124.1 | - | 1 - 2 | 562 | 75 |