Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL26
Entrez Gene ID
55801
Gene Name
interleukin 26
Gene Aliases
AK155, IL-26
Chromosome Location
Chr.12: 68201349 - 68225791 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018402.1 NP_060872.1 3 - 4 395 81
GenBank mRNA AJ251549.1 - 3 - 4 395 81
BC066270.1 - 3 - 4 394 81
BC066271.1 - 3 - 4 394 81

View Full Product Details