The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282529.1 | NP_001269458.1 | 20 - 21 | 2592 | 76 |
NM_001282530.1 | NP_001269459.1 | 8 - 9 | 1135 | 76 | |
NM_018449.3 | NP_060919.3 | 24 - 25 | 2906 | 76 | |
GenBank mRNA | AB040924.1 | - | 12 - 13 | 1708 | 76 |
AF157327.1 | - | 10 - 11 | 1121 | 76 | |
AK025357.1 | - | 5 - 6 | 548 | 76 | |
AK025801.1 | - | 13 - 14 | 1796 | 76 | |
AK026088.1 | - | 16 - 17 | 2074 | 76 | |
AK057564.1 | - | 5 - 6 | 1461 | 76 | |
AK074884.1 | - | 11 - 12 | 1265 | 76 | |
AK125935.1 | - | 7 - 8 | 2390 | 76 | |
AK299183.1 | - | 18 - 19 | 2314 | 76 | |
AK302754.1 | - | 23 - 24 | 2774 | 76 | |
AK303493.1 | - | 20 - 21 | 2530 | 76 | |
BC007890.2 | - | 2 - 3 | 225 | 76 | |
BC037416.1 | - | 5 - 6 | 539 | 76 |