Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACTR10
Entrez Gene ID
55860
Gene Name
actin-related protein 10 homolog
Gene Aliases
ACTR11, Arp10, Arp11, HARP11
Chromosome Location
Chr.14: 58200115 - 58235636 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018477.2 NP_060947.1 8 - 9 734 68
XM_011536963.2 XP_011535265.1 6 - 7 514 68
GenBank mRNA AF220190.1 - 8 - 9 711 68
AK000544.1 - 8 - 9 714 68
AK315537.1 - 8 - 9 679 68
BC005208.1 - 8 - 9 695 68
BC011997.1 - 8 - 9 668 68
BX161491.1 - 3 - 4 778 68
BX248282.1 - 8 - 9 630 68

View Full Product Details