The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018518.4 | NP_060988.3 | 16 - 17 | 2367 | 59 |
NM_182751.2 | NP_877428.1 | 16 - 17 | 2370 | 59 | |
XM_011519538.2 | XP_011517840.1 | 16 - 17 | 2340 | 59 | |
GenBank mRNA | AB042719.1 | - | 16 - 17 | 2334 | 59 |
AF119869.1 | - | 5 - 6 | 706 | 59 | |
AK292701.1 | - | 16 - 17 | 2345 | 59 | |
AL136840.1 | - | 15 - 16 | 2333 | 59 | |
BC004876.2 | - | 9 - 10 | 1285 | 59 | |
BC009108.1 | - | 11 - 12 | 1585 | 59 | |
BC101727.1 | - | 16 - 17 | 2339 | 59 | |
BC143490.1 | - | 16 - 17 | 2332 | 59 |