The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018649.2 | NP_061119.1 | 1 - 2 | 399 | 101 |
GenBank mRNA | AF151534.1 | - | 1 - 2 | 153 | 101 |
AF336304.1 | - | 1 - 2 | 131 | 101 | |
AK022475.1 | - | 1 - 2 | 123 | 101 | |
AK022776.1 | - | 1 - 2 | 154 | 101 | |
AK131057.1 | - | 1 - 2 | 150 | 101 | |
BC016172.2 | - | 1 - 2 | 155 | 101 | |
CX752382.1 | - | 1 - 2 | 399 | 101 |