Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLGA6B
Entrez Gene ID
55889
Gene Name
golgin A6 family member B
Gene Aliases
GOLGA, GOLGA6D
Chromosome Location
Chr.15: 72638291 - 72669604 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018652.4 NP_061122.4 13 - 14 1547 124
XM_017022406.1 XP_016877895.1 18 - 19 2435 124

Gene Symbol
GOLGA6A
Entrez Gene ID
342096
Gene Name
golgin A6 family member A
Gene Aliases
GLP, GOLGA6
Chromosome Location
Chr.15: 74069857 - 74082550 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038640.2 NP_001033729.2 13 - 14 124
GenBank mRNA AF263742.1 - 13 - 14 124
AK292574.1 - 13 - 14 124
BC131515.1 - 13 - 14 124

Gene Symbol
GOLGA6C
Entrez Gene ID
653641
Gene Name
golgin A6 family member C
Gene Aliases
-
Chromosome Location
Chr.15: 75258404 - 75273455 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164404.1 NP_001157876.1 13 - 14 124
XM_017022519.1 XP_016878008.1 13 - 14 124

Gene Symbol
GOLGA6D
Entrez Gene ID
653643
Gene Name
golgin A6 family member D
Gene Aliases
-
Chromosome Location
Chr.15: 75282841 - 75295807 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145224.1 NP_001138696.1 13 - 14 124

View Full Product Details