Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BIN3
Entrez Gene ID
55909
Gene Name
bridging integrator 3
Gene Aliases
-
Chromosome Location
Chr.8: 22620418 - 22669148 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018688.4 NP_061158.1 4 - 5 261 113
XM_011544587.1 XP_011542889.1 4 - 5 261 113
GenBank mRNA AF271732.1 - 4 - 5 224 113
AK023980.1 - 4 - 5 208 113
AK222470.1 - 4 - 5 233 113
BC001223.1 - 3 - 4 145 113
BC009824.2 - 4 - 5 195 113

View Full Product Details