The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018948.3 | NP_061821.1 | 3 - 4 | 454 | 57 |
XM_005263477.2 | XP_005263534.1 | 3 - 4 | 468 | 57 | |
XM_006710697.2 | XP_006710760.1 | 3 - 4 | 1638 | 57 | |
GenBank mRNA | AJ276373.1 | - | 2 - 3 | 205 | 57 |
AK096149.1 | - | 3 - 4 | 429 | 57 | |
AK315718.1 | - | 3 - 4 | 409 | 57 | |
BC025337.1 | - | 3 - 4 | 417 | 57 | |
DB033666.1 | - | 3 - 4 | 479 | 57 |