Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC93
Entrez Gene ID
54520
Gene Name
coiled-coil domain containing 93
Gene Aliases
-
Chromosome Location
Chr.2: 117915478 - 118014163 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019044.4 NP_061917.3 23 - 24 2015 100
XM_011511359.1 XP_011509661.1 23 - 24 2012 100
XM_011511361.1 XP_011509663.1 22 - 23 1769 100
XM_011511362.1 XP_011509664.1 22 - 23 1883 100
GenBank mRNA AB209993.1 - 23 - 24 1934 100
AK001858.1 - 11 - 12 809 100
AK025611.1 - 22 - 23 1783 100
AK290722.1 - 23 - 24 1922 100
BC028609.1 - 23 - 24 1988 100
BC093018.1 - 23 - 24 1905 100

View Full Product Details