The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306188.1 | NP_001293117.1 | 3 - 4 | 499 | 69 |
NM_019556.2 | NP_062456.1 | 3 - 4 | 499 | 69 | |
XM_011531366.2 | XP_011529668.1 | 3 - 4 | 453 | 69 | |
XM_017029653.1 | XP_016885142.1 | 4 - 5 | 692 | 69 | |
XM_017029654.1 | XP_016885143.1 | 3 - 4 | 453 | 69 | |
GenBank mRNA | AK299074.1 | - | 3 - 4 | 412 | 69 |
AK299172.1 | - | 2 - 3 | 289 | 69 | |
AK316570.1 | - | 3 - 4 | 374 | 69 | |
AL137163.1 | - | 3 - 4 | 422 | 69 | |
BC005700.1 | - | 3 - 4 | 402 | 69 | |
CB997365.1 | - | 3 - 4 | 407 | 69 | |
CN428237.1 | - | 3 - 4 | 501 | 69 | |
DQ323992.1 | - | 2 - 3 | 235 | 69 |