Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEPCE
Entrez Gene ID
56257
Gene Name
methylphosphate capping enzyme
Gene Aliases
BCDIN3
Chromosome Location
Chr.7: 100428790 - 100434126 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194990.1 NP_001181919.1 2 - 3 1322 66
NM_001194991.1 NP_001181920.1 2 - 3 907 66
NM_001194992.1 NP_001181921.1 2 - 3 772 66
NM_019606.5 NP_062552.2 1 - 2 2055 66
XM_011516410.2 XP_011514712.1 2 - 3 1380 66
GenBank mRNA AK000264.1 - 1 - 2 710 66
AK055964.1 - 2 - 3 1295 66
AK291366.1 - 1 - 2 1850 66
BC000556.2 - 1 - 2 1780 66
BC016396.1 - 1 - 2 2055 66
BC018935.2 - 1 - 2 1780 66

View Full Product Details