Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TRIM49
- Entrez Gene ID
-
57093
- Gene Name
- tripartite motif containing 49
- Gene Aliases
-
RNF18, TRIM49A, TRIM49L2
- Chromosome Location
-
Chr.11: 89797655 - 89808575 on Build GRCh38
- Gene Symbol
- TRIM49B
- Entrez Gene ID
-
283116
- Gene Name
- tripartite motif containing 49B
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 49031596 - 49038049 on Build GRCh38
- Gene Symbol
- TRIM49C
- Entrez Gene ID
-
642612
- Gene Name
- tripartite motif containing 49C
- Gene Aliases
-
TRIM49L2
- Chromosome Location
-
Chr.11: 90031106 - 90042025 on Build GRCh38
View Full Product Details