The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020366.3 | NP_065099.3 | 10 - 11 | 1310 | 83 |
XM_005267879.2 | XP_005267936.1 | 1 - 2 | 302 | 83 | |
XM_005267880.2 | XP_005267937.1 | 1 - 2 | 302 | 83 | |
XM_011536978.1 | XP_011535280.1 | 1 - 2 | 302 | 83 | |
XM_011536979.1 | XP_011535281.1 | 1 - 2 | 302 | 83 | |
XM_011536980.1 | XP_011535282.1 | 1 - 2 | 302 | 83 | |
XM_011536981.1 | XP_011535283.1 | 1 - 2 | 302 | 83 | |
XM_011536982.1 | XP_011535284.1 | 1 - 2 | 302 | 83 | |
XM_017021473.1 | XP_016876962.1 | 1 - 2 | 302 | 83 | |
GenBank mRNA | AF260257.1 | - | 1 - 2 | 262 | 83 |
AJ417067.1 | - | 10 - 11 | 1310 | 83 | |
AK301780.1 | - | 1 - 2 | 316 | 83 | |
BC039089.1 | - | 1 - 2 | 301 | 83 |