The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020367.5 | NP_065100.2 | 3 - 4 | 488 | 81 |
NR_104461.1 | 3 - 4 | 488 | 81 | ||
XM_005253711.4 | XP_005253768.1 | 3 - 4 | 882 | 81 | |
XM_011520970.1 | XP_011519272.1 | 4 - 5 | 604 | 81 | |
XM_011520971.2 | XP_011519273.1 | 4 - 5 | 1004 | 81 | |
XM_011520973.2 | XP_011519275.1 | 3 - 4 | 317 | 81 | |
XM_017019670.1 | XP_016875159.1 | 4 - 5 | 526 | 81 | |
GenBank mRNA | AF263540.1 | - | 2 - 3 | 303 | 81 |
AK314916.1 | - | 3 - 4 | 389 | 81 | |
BC017569.1 | - | 3 - 4 | 414 | 81 | |
BC031073.1 | - | 3 - 4 | 476 | 81 |