The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020651.3 | NP_065702.2 | 3 - 4 | 657 | 70 |
XM_011532994.2 | XP_011531296.1 | 3 - 4 | 1316 | 70 | |
XM_011532996.2 | XP_011531298.1 | 3 - 4 | 2075 | 70 | |
XM_017004520.1 | XP_016860009.1 | 4 - 5 | 791 | 70 | |
GenBank mRNA | AF300987.1 | - | 2 - 3 | 212 | 70 |
AF302505.1 | - | 2 - 3 | 4235 | 70 | |
AJ278859.1 | - | 2 - 3 | 197 | 70 | |
AK312441.1 | - | 3 - 4 | 606 | 70 | |
AY364257.1 | - | 2 - 3 | 197 | 70 | |
BC050019.1 | - | 3 - 4 | 611 | 70 | |
BC050533.1 | - | 3 - 4 | 593 | 70 | |
BC063611.1 | - | 3 - 4 | 585 | 70 |