The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134771.1 | NP_001128243.1 | 6 - 7 | 760 | 80 |
NM_020708.4 | NP_065759.1 | 6 - 7 | 778 | 80 | |
XM_017027981.1 | XP_016883470.1 | 6 - 7 | 760 | 80 | |
GenBank mRNA | AB033002.1 | - | 6 - 7 | 593 | 80 |
AF208159.1 | - | 6 - 7 | 615 | 80 | |
AK098371.1 | - | 6 - 7 | 764 | 80 | |
AK289758.1 | - | 6 - 7 | 737 | 80 | |
BC132668.1 | - | 6 - 7 | 703 | 80 | |
BC132670.1 | - | 6 - 7 | 703 | 80 | |
KC295694.1 | - | 5 - 6 | 1139 | 80 | |
KC295701.1 | - | 6 - 7 | 778 | 80 |