The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128432.2 | NP_001121904.1 | 1 - 2 | 162 | 82 |
NM_020973.4 | NP_066024.1 | 1 - 2 | 162 | 82 | |
NR_102355.1 | 1 - 2 | 162 | 82 | ||
NR_102356.1 | 1 - 2 | 162 | 82 | ||
GenBank mRNA | AB017913.1 | - | 1 - 2 | 111 | 82 |
AF317840.1 | - | 1 - 2 | 60 | 82 | |
AF323990.1 | - | 1 - 2 | 109 | 82 | |
AJ278964.1 | - | 1 - 2 | 60 | 82 | |
AK222963.1 | - | 1 - 2 | 109 | 82 | |
AK292746.1 | - | 1 - 2 | 109 | 82 | |
AK313189.1 | - | 1 - 2 | 114 | 82 | |
BC029362.1 | - | 1 - 2 | 151 | 82 | |
BC101829.1 | - | 1 - 2 | 97 | 82 | |
BC109377.1 | - | 1 - 2 | 142 | 82 | |
BC136264.1 | - | 1 - 2 | 97 | 82 | |
BP383134.1 | - | - | 162 | 82 |