Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GINS1
Entrez Gene ID
9837
Gene Name
GINS complex subunit 1
Gene Aliases
PSF1
Chromosome Location
Chr.20: 25407470 - 25448568 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021067.4 NP_066545.3 1 - 2 215 87
NR_134574.1 1 - 2 215 87
GenBank mRNA AK294593.1 - 1 - 2 169 87
BC012542.1 - 1 - 2 216 87
BP290581.1 - - 215 87
D80008.1 - 1 - 2 171 87

View Full Product Details