The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316345.1 | NP_001303274.1 | 17 - 18 | 2108 | 76 |
NM_021111.2 | NP_066934.1 | 15 - 16 | 1972 | 76 | |
XM_017015207.1 | XP_016870696.1 | 16 - 17 | 2008 | 76 | |
XM_017015208.1 | XP_016870697.1 | 16 - 17 | 2082 | 76 | |
XM_017015209.1 | XP_016870698.1 | 16 - 17 | 2071 | 76 | |
XM_017015210.1 | XP_016870699.1 | 14 - 15 | 1823 | 76 | |
XM_017015211.1 | XP_016870700.1 | 15 - 16 | 1974 | 76 | |
GenBank mRNA | D50406.1 | - | 15 - 16 | 1995 | 76 |