The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021629.3 | NP_067642.1 | 4 - 5 | 486 | 78 |
XM_005247692.2 | XP_005247749.1 | 4 - 5 | 407 | 78 | |
XM_006713721.2 | XP_006713784.1 | 4 - 5 | 417 | 78 | |
GenBank mRNA | AF300648.1 | - | 3 - 4 | 206 | 78 |
AK001890.1 | - | 4 - 5 | 486 | 78 | |
AK290571.1 | - | 4 - 5 | 417 | 78 | |
AK297174.1 | - | 4 - 5 | 380 | 78 | |
BC000873.1 | - | 4 - 5 | 296 | 78 |