Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SVIL
Entrez Gene ID
6840
Gene Name
supervillin
Gene Aliases
-
Chromosome Location
Chr.10: 29457338 - 29737001 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001323599.1 NP_001310528.1 13 - 14 2033 99
NM_001323600.1 NP_001310529.1 11 - 12 1634 99
NM_021738.2 NP_068506.2 12 - 13 3000 99
XM_005252570.2 XP_005252627.1 14 - 15 2897 99
XM_005252571.3 XP_005252628.1 14 - 15 2823 99
XM_005252573.2 XP_005252630.1 12 - 13 3029 99
XM_011519630.2 XP_011517932.2 12 - 13 3742 99
XM_011519632.2 XP_011517934.2 11 - 12 3571 99
XM_011519634.1 XP_011517936.1 14 - 15 2955 99
XM_011519635.1 XP_011517937.1 12 - 13 2922 99
XM_011519636.2 XP_011517938.2 11 - 12 2812 99
XM_011519637.2 XP_011517939.2 9 - 10 2560 99
XM_017016575.1 XP_016872064.1 12 - 13 3742 99
XM_017016576.1 XP_016872065.1 11 - 12 3661 99
XM_017016577.1 XP_016872066.1 12 - 13 3742 99
XM_017016579.1 XP_016872068.1 11 - 12 2812 99
XM_017016580.1 XP_016872069.1 10 - 11 2641 99
XM_017016581.1 XP_016872070.1 9 - 10 2560 99
XM_017016583.1 XP_016872072.1 12 - 13 3742 99
GenBank mRNA AF109135.1 - 12 - 13 3000 99

View Full Product Details