The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135053.1 | NP_001128525.1 | 8 - 9 | 1138 | 78 |
NM_001135054.1 | NP_001128526.1 | 8 - 9 | 1211 | 78 | |
NM_021805.2 | NP_068577.2 | 8 - 9 | 1107 | 78 | |
XM_005253044.2 | XP_005253101.1 | 8 - 9 | 1211 | 78 | |
XM_005253045.1 | XP_005253102.1 | 8 - 9 | 1107 | 78 | |
XM_005253046.1 | XP_005253103.1 | 8 - 9 | 1195 | 78 | |
XM_005253048.4 | XP_005253105.1 | 6 - 7 | 901 | 78 | |
XM_017018099.1 | XP_016873588.1 | 8 - 9 | 1211 | 78 | |
XM_017018100.1 | XP_016873589.1 | 7 - 8 | 1040 | 78 | |
GenBank mRNA | AK025099.1 | - | 8 - 9 | 1107 | 78 |
AK172830.1 | - | 8 - 9 | 1107 | 78 | |
AY358342.1 | - | 7 - 8 | 984 | 78 | |
BC003591.1 | - | 7 - 8 | 1037 | 78 | |
BC025953.1 | - | 8 - 9 | 1066 | 78 | |
BC106007.1 | - | 8 - 9 | 1188 | 78 | |
BX369506.2 | - | 3 - 4 | 306 | 78 | |
CR457338.1 | - | 6 - 7 | 881 | 78 |