The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021922.2 | NP_068741.1 | 8 - 9 | 1566 | 79 |
XM_005248885.2 | XP_005248942.1 | 8 - 9 | 1567 | 79 | |
XM_005248886.2 | XP_005248943.1 | 7 - 8 | 1519 | 79 | |
XM_011514343.2 | XP_011512645.1 | 8 - 9 | 1281 | 79 | |
GenBank mRNA | AF265210.1 | - | 8 - 9 | 1566 | 79 |
AK292522.1 | - | 8 - 9 | 1577 | 79 | |
AK312617.1 | - | 8 - 9 | 1533 | 79 | |
BC046359.1 | - | 8 - 9 | 1572 | 79 |