Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKNOX2
Entrez Gene ID
63876
Gene Name
PBX/knotted 1 homeobox 2
Gene Aliases
PREP2
Chromosome Location
Chr.11: 125111997 - 125433389 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022062.2 NP_071345.2 7 - 8 904 68
XM_005271642.2 XP_005271699.1 6 - 7 793 68
XM_005271643.2 XP_005271700.1 4 - 5 610 68
XM_006718894.2 XP_006718957.1 6 - 7 680 68
XM_011542944.2 XP_011541246.1 7 - 8 989 68
XM_011542945.2 XP_011541247.1 8 - 9 949 68
XM_011542947.2 XP_011541249.1 5 - 6 558 68
XM_017018110.1 XP_016873599.1 7 - 8 3739 68
XM_017018111.1 XP_016873600.1 5 - 6 599 68
GenBank mRNA AB065001.1 - 4 - 5 582 68
AJ417081.1 - 4 - 5 554 68
AK023136.1 - 4 - 5 582 68
AK295857.1 - 6 - 7 771 68
AK298999.1 - 6 - 7 673 68
AK316001.1 - 6 - 7 704 68
AK316268.1 - 5 - 6 692 68
BX648615.1 - 5 - 6 681 68

View Full Product Details