Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMSN1
Entrez Gene ID
64092
Gene Name
SAM domain, SH3 domain and nuclear localization signals 1
Gene Aliases
HACS1, NASH1, SASH2, SH3D6B, SLy2
Chromosome Location
Chr.21: 14485228 - 14583402 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256370.1 NP_001243299.1 3 - 4 510 90
NM_001286523.1 NP_001273452.1 3 - 4 354 90
NM_022136.4 NP_071419.3 2 - 3 230 90
XM_011529684.2 XP_011527986.1 3 - 4 270 90
XM_011529686.1 XP_011527988.1 2 - 3 227 90
XM_017028427.1 XP_016883916.1 2 - 3 227 90
GenBank mRNA AF218085.2 - 3 - 4 510 90
AF222927.1 - 2 - 3 213 90
AK125144.1 - 2 - 3 213 90
BC029112.1 - 2 - 3 204 90
DA819791.1 - 2 - 3 230 90
HG380513.1 - 3 - 4 260 90
HG380514.1 - 3 - 4 329 90

View Full Product Details