Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CARD9
Entrez Gene ID
64170
Gene Name
caspase recruitment domain family member 9
Gene Aliases
CANDF2, hCARD9
Chromosome Location
Chr.9: 136363956 - 136373681 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK024001.1 - 7 - 8 1229 112
AK127481.1 - 2 - 3 1011 112

View Full Product Details