Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- IKZF4
- Entrez Gene ID
-
64375
- Gene Name
- IKAROS family zinc finger 4
- Gene Aliases
-
EOS, ZNFN1A4
- Chromosome Location
-
Chr.12: 56007647 - 56038435 on Build GRCh38
View Full Product Details