Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IKZF5
Entrez Gene ID
64376
Gene Name
IKAROS family zinc finger 5
Gene Aliases
PEGASUS, ZNFN1A5
Chromosome Location
Chr.10: 122990806 - 123008850 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271840.1 NP_001258769.1 3 - 4 442 121
NR_073475.1 3 - 4 486 121
NR_073477.1 3 - 4 473 121
NR_073478.1 2 - 3 335 121
NR_073479.1 4 - 5 594 121
XM_006717947.3 XP_006718010.1 4 - 5 556 121
XM_017016550.1 XP_016872039.1 3 - 4 434 121
XM_017016551.1 XP_016872040.1 4 - 5 530 121
XM_017016552.1 XP_016872041.1 3 - 4 422 121
GenBank mRNA AF230808.1 - 1 - 2 132 121
AK026626.1 - 3 - 4 431 121
AK122899.1 - 4 - 5 2233 121
BC022564.1 - 3 - 4 441 121
BX647958.1 - 3 - 4 435 121
CR749800.1 - 3 - 4 428 121
DA218215.1 - 3 - 4 406 121
DA227834.1 - 2 - 3 290 121
DB080324.1 - 3 - 4 473 121

View Full Product Details