The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029991.1 | NP_001025162.1 | 6 - 7 | 804 | 60 |
NM_022734.2 | NP_073571.1 | 6 - 7 | 804 | 60 | |
XM_006720236.2 | XP_006720299.1 | 5 - 6 | 564 | 60 | |
XM_017021601.1 | XP_016877090.1 | 5 - 6 | 564 | 60 | |
GenBank mRNA | AF321002.1 | - | 6 - 7 | 639 | 60 |
AK024512.1 | - | 6 - 7 | 630 | 60 | |
BC005053.1 | - | 6 - 7 | 623 | 60 | |
KU178747.1 | - | 6 - 7 | 607 | 60 | |
KU178748.1 | - | 6 - 7 | 639 | 60 | |
KU178749.1 | - | 6 - 7 | 607 | 60 |