Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXO5
Entrez Gene ID
64789
Gene Name
exonuclease 5
Gene Aliases
C1orf176, DEM1, Exo V, hExo5
Chromosome Location
Chr.1: 40508719 - 40516556 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022774.1 NP_073611.1 2 - 3 180 67
XM_005271125.3 XP_005271182.1 3 - 4 428 67
XM_005271126.3 XP_005271183.1 2 - 3 240 67
XM_017002099.1 XP_016857588.1 3 - 4 852 67
XM_017002100.1 XP_016857589.1 3 - 4 407 67
XM_017002101.1 XP_016857590.1 3 - 4 378 67
XM_017002102.1 XP_016857591.1 2 - 3 780 67
XM_017002103.1 XP_016857592.1 2 - 3 206 67
GenBank mRNA AK023245.1 - 1 - 2 1007 67
AK024797.1 - 2 - 3 180 67
AK094092.1 - 2 - 3 1002 67
BC021969.2 - 3 - 4 341 67

View Full Product Details