The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022901.2 | NP_075052.1 | 4 - 5 | 890 | 99 |
GenBank mRNA | AK024955.1 | - | 4 - 5 | 869 | 99 |
AK222987.1 | - | 4 - 5 | 862 | 99 | |
BC056252.1 | - | 4 - 5 | 894 | 99 | |
BC126156.1 | - | 4 - 5 | 849 | 99 | |
BC136298.1 | - | 4 - 5 | 849 | 99 |