The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024012.3 | NP_076917.1 | 1 - 2 | 1316 | 82 |
GenBank mRNA | AF498985.1 | - | 1 - 2 | 740 | 82 |
AK094255.1 | - | 1 - 2 | 1316 | 82 | |
BC112021.1 | - | 1 - 2 | 804 | 82 | |
BC113468.1 | - | 1 - 2 | 804 | 82 |