The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024038.2 | NP_076943.1 | 1 - 2 | 406 | 67 |
GenBank mRNA | AK027588.1 | - | 1 - 2 | 301 | 67 |
BC000216.2 | - | 1 - 2 | 406 | 67 | |
BC004533.2 | - | 1 - 2 | 351 | 67 | |
BC015347.1 | - | 1 - 2 | 365 | 67 |