The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301011.1 | NP_001287940.1 | 2 - 3 | 506 | 81 |
NM_024296.4 | NP_077272.2 | 2 - 3 | 506 | 81 | |
XM_011542113.2 | XP_011540415.1 | 2 - 3 | 1265 | 81 | |
XM_017002306.1 | XP_016857795.1 | 2 - 3 | 506 | 81 | |
XM_017002307.1 | XP_016857796.1 | 2 - 3 | 1260 | 81 | |
XM_017002308.1 | XP_016857797.1 | 2 - 3 | 506 | 81 | |
XM_017002309.1 | XP_016857798.1 | 2 - 3 | 1263 | 81 | |
GenBank mRNA | AK225113.1 | - | 2 - 3 | 276 | 81 |
AK291904.1 | - | 2 - 3 | 516 | 81 | |
BC022848.2 | - | 2 - 3 | 240 | 81 |