The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024319.3 | NP_077295.1 | 2 - 3 | 483 | 89 |
NR_130975.1 | 2 - 3 | 483 | 89 | ||
GenBank mRNA | AK123377.1 | - | 2 - 3 | 463 | 89 |
AK126087.1 | - | 2 - 3 | 2432 | 89 | |
AY137773.1 | - | 2 - 3 | 344 | 89 | |
BC002843.2 | - | 2 - 3 | 342 | 89 | |
BC062585.1 | - | 2 - 3 | 339 | 89 |