Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRDM14
Entrez Gene ID
63978
Gene Name
PR domain 14
Gene Aliases
PFM11
Chromosome Location
Chr.8: 70051126 - 70071327 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024504.3 NP_078780.1 5 - 6 1386 102
XM_011517572.1 XP_011515874.1 4 - 5 666 102
GenBank mRNA AF319458.1 - 5 - 6 1386 102
AK022595.1 - 5 - 6 1386 102
BC052311.1 - 5 - 6 1299 102
KJ463837.1 - 4 - 5 1184 102

View Full Product Details