Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SPAG16
- Entrez Gene ID
-
79582
- Gene Name
- sperm associated antigen 16
- Gene Aliases
-
PF20, WDR29
- Chromosome Location
-
Chr.2: 213284369 - 214410501 on Build GRCh38
View Full Product Details