The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024560.2 | NP_078836.1 | 9 - 10 | 1448 | 105 |
XM_005269150.2 | XP_005269207.1 | 9 - 10 | 1382 | 105 | |
XM_011538740.2 | XP_011537042.1 | 9 - 10 | 1097 | 105 | |
GenBank mRNA | AK025616.1 | - | 9 - 10 | 1399 | 105 |
AX764480.1 | - | 9 - 10 | 1598 | 105 | |
AX764482.1 | - | 9 - 10 | 1354 | 105 | |
BC009317.2 | - | 9 - 10 | 1448 | 105 | |
BC015769.1 | - | 9 - 10 | 1448 | 105 |