The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193528.1 | NP_001180457.1 | 6 - 7 | 923 | 76 |
NM_024583.4 | NP_078859.2 | 6 - 7 | 1014 | 76 | |
XM_005246853.2 | XP_005246910.1 | 6 - 7 | 1419 | 76 | |
XM_005246854.4 | XP_005246911.1 | 6 - 7 | 2579 | 76 | |
XM_005246855.2 | XP_005246912.1 | 6 - 7 | 1467 | 76 | |
XM_005246856.2 | XP_005246913.1 | 6 - 7 | 1423 | 76 | |
XM_005246857.2 | XP_005246914.1 | 6 - 7 | 1123 | 76 | |
XM_011511839.2 | XP_011510141.1 | 5 - 6 | 823 | 76 | |
XM_017004909.1 | XP_016860398.1 | 6 - 7 | 2011 | 76 | |
XM_017004910.1 | XP_016860399.1 | 6 - 7 | 1489 | 76 | |
GenBank mRNA | AF279776.1 | - | 2 - 3 | 351 | 76 |
AK026795.1 | - | 2 - 3 | 361 | 76 | |
AK057912.1 | - | 6 - 7 | 992 | 76 | |
AK295357.1 | - | 6 - 7 | 907 | 76 | |
AK296558.1 | - | 5 - 6 | 813 | 76 | |
BC031821.1 | - | 3 - 4 | 475 | 76 | |
BC119684.1 | - | 6 - 7 | 965 | 76 | |
BC119685.1 | - | 6 - 7 | 965 | 76 |