Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD55
Entrez Gene ID
79722
Gene Name
ankyrin repeat domain 55
Gene Aliases
-
Chromosome Location
Chr.5: 56099680 - 56233359 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024669.2 NP_078945.2 10 - 11 1785 78
XM_011543646.1 XP_011541948.1 4 - 5 1099 78
XM_017009852.1 XP_016865341.1 10 - 11 1697 78
XM_017009853.1 XP_016865342.1 9 - 10 1669 78
XM_017009854.1 XP_016865343.1 7 - 8 3019 78
GenBank mRNA AK021857.1 - 2 - 3 838 78
AK123389.1 - 10 - 11 1785 78
BC106914.2 - 2 - 3 1005 78
BC106915.2 - 2 - 3 78

View Full Product Details