The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024734.3 | NP_079010.2 | 9 - 10 | 2628 | 66 |
XM_011537158.1 | XP_011535460.1 | 9 - 10 | 2624 | 66 | |
XM_011537159.2 | XP_011535461.1 | 9 - 10 | 2624 | 66 | |
XM_017021646.1 | XP_016877135.1 | 9 - 10 | 2794 | 66 | |
XM_017021647.1 | XP_016877136.1 | 9 - 10 | 2624 | 66 | |
GenBank mRNA | AB033014.1 | - | 1 - 2 | 944 | 66 |
AB047979.1 | - | 9 - 10 | 2585 | 66 | |
AK022279.1 | - | 4 - 5 | 1972 | 66 | |
AK023828.1 | - | 1 - 2 | 1594 | 66 | |
AK025226.1 | - | 9 - 10 | 2571 | 66 | |
AK314315.1 | - | 9 - 10 | 2626 | 66 | |
AL117526.1 | - | 9 - 10 | 2480 | 66 | |
BC014554.2 | - | 1 - 2 | 1168 | 66 | |
BC025251.1 | - | 1 - 2 | 1567 | 66 | |
BC032699.1 | - | 4 - 5 | 1947 | 66 | |
BC068482.1 | - | 9 - 10 | 2577 | 66 |