The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031720.3 | NP_001026890.2 | 4 - 5 | 1221 | 104 |
NM_024751.3 | NP_079027.2 | 4 - 5 | 1142 | 104 | |
XM_005263222.3 | XP_005263279.1 | 4 - 5 | 1409 | 104 | |
XM_011532248.2 | XP_011530550.1 | 4 - 5 | 1365 | 104 | |
XM_011532249.2 | XP_011530551.1 | 4 - 5 | 1227 | 104 | |
XM_011532252.2 | XP_011530554.1 | 4 - 5 | 1599 | 104 | |
GenBank mRNA | AK023335.1 | - | 4 - 5 | 1400 | 104 |
AK225563.1 | - | 4 - 5 | 1400 | 104 | |
AK292355.1 | - | 4 - 5 | 1224 | 104 | |
AK303547.1 | - | 2 - 3 | 488 | 104 | |
BC032942.2 | - | 4 - 5 | 1218 | 104 | |
BX648355.1 | - | 4 - 5 | 1143 | 104 |