Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTCD2
Entrez Gene ID
79810
Gene Name
pentatricopeptide repeat domain 2
Gene Aliases
-
Chromosome Location
Chr.5: 72320367 - 72368395 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK022660.1 - 8 - 9 1339 93
BC018720.1 - 10 - 11 1553 93

View Full Product Details