The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300800.1 | NP_001287729.1 | 3 - 4 | 356 | 71 |
NM_024771.3 | NP_079047.2 | 3 - 4 | 285 | 71 | |
XM_006718689.3 | XP_006718752.1 | 3 - 4 | 272 | 71 | |
XM_011545254.2 | XP_011543556.1 | 4 - 5 | 359 | 71 | |
XM_011545255.2 | XP_011543557.1 | 4 - 5 | 359 | 71 | |
GenBank mRNA | AK023910.1 | - | 3 - 4 | 254 | 71 |
AK299041.1 | - | 3 - 4 | 350 | 71 | |
BC041617.1 | - | 3 - 4 | 214 | 71 | |
BC052298.1 | - | 3 - 4 | 214 | 71 | |
BX647309.1 | - | 2 - 3 | 1311 | 71 | |
DA649381.1 | - | 3 - 4 | 290 | 71 | |
DC307984.1 | - | 3 - 4 | 439 | 71 | |
HY009487.1 | - | 3 - 4 | 356 | 71 | |
JF263566.1 | - | 3 - 4 | 254 | 71 |