The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015041.2 | NP_055856.1 | 6 - 7 | 701 | 98 |
NM_024793.2 | NP_079069.1 | 2 - 3 | 159 | 98 | |
XM_006720867.3 | XP_006720930.1 | 5 - 6 | 552 | 98 | |
XM_017023068.1 | XP_016878557.1 | 6 - 7 | 716 | 98 | |
XM_017023069.1 | XP_016878558.1 | 6 - 7 | 716 | 98 | |
XM_017023070.1 | XP_016878559.1 | 6 - 7 | 701 | 98 | |
XM_017023071.1 | XP_016878560.1 | 6 - 7 | 1690 | 98 | |
XM_017023072.1 | XP_016878561.1 | 6 - 7 | 1690 | 98 | |
XM_017023073.1 | XP_016878562.1 | 5 - 6 | 552 | 98 | |
GenBank mRNA | AB014543.1 | - | 6 - 7 | 628 | 98 |
AB089691.1 | - | 6 - 7 | 683 | 98 | |
AK023359.1 | - | 2 - 3 | 159 | 98 | |
BC017070.2 | - | 6 - 7 | 629 | 98 |