Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DENND1A
- Entrez Gene ID
-
57706
- Gene Name
- DENN domain containing 1A
- Gene Aliases
-
FAM31A, KIAA1608
- Chromosome Location
-
Chr.9: 123379654 - 123930158 on Build GRCh38
View Full Product Details